4,4,5,5,6,6,6-Heptafluoro-1-hexanol ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4,5,5,6,6,6-Heptafluorohexan-1-ol

008982-1G 008982-5G
101796-012EA 41.54 USD
101796-012 101796-014
4,4,5,5,6,6,6-Heptafluoro-1-hexanol ≥97%
4,4,5,5,6,6,6-Heptafluoro-1-hexanol

Hazard code(s): Irritant

Formula: C₆H₇F₇O
MW: 228.11 g/mol
Boiling Pt: 140…141 °C (745mm)
MDL Number: MFCD00077554
CAS Number: 679-02-7

Order Now

Specification Test Results

Boiling point 140-141°C/7

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR