4-Fluoro-α-methylbenzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: p-Fluoro-α-methylbenzyl alcohol, 1-(4-Fluorophenyl)ethyl Alcohol, 1-(4-Fluorophenyl)ethanol, 4-Fluorophenyl methyl carbinol

008966-5G 008966-25G
101795-964EA 21.54 USD
101795-964 101795-966
4-Fluoro-α-methylbenzyl alcohol ≥97% for synthesis
4-Fluoro-α-methylbenzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₉FO
MW: 140.16
Boiling Pt: 49.5
Melting Pt: 9.3
Density: 1.120 g/cm³
MDL Number: MFCD00004515
CAS Number: 403-41-8
EINECS: 206-959-3

Order Now

Specification Test Results

Boiling point 91°C/10mm
Density 1.12
ND20 1.501


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR