3-Fluoro-α-methylbenzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific

008965-1G 008965-5G
101795-960EA 33.85 USD
101795-960 101795-962
3-Fluoro-α-methylbenzyl alcohol ≥97% for synthesis
3-Fluoro-α-methylbenzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₉FO
MW: 140.16
MDL Number: MFCD00014402
CAS Number: 402-63-1
EINECS: 206-950-4

Order Now

Specification Test Results

ND20 1.505


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR