2-Fluoro-α-methylbenzyl alcohol ≥97%

Supplier: Matrix Scientific

008964-1G 008964-5G
101795-956EA 61.54 USD
101795-956 101795-958
2-Fluoro-α-methylbenzyl alcohol ≥97%
2-Fluoro-α-methylbenzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 110 °C (135mm)
MDL Number: MFCD00014401
CAS Number: 445-26-1
EINECS: 207-155-5

Order Now

Specification Test Results

Boiling point 110°C/135mm
ND20 1.505

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR