(4-Fluorophenoxy)acetic acid
Supplier: Matrix Scientific
Synonyms:
4-Fluorophenoxyacetic Acid
008961-25G
008961-100G
101795-944EA
29.23
USD
101795-944
101795-946
(4-Fluorophenoxy)acetic acid
(4-Fluorophenoxy)acetic acid
Hazard code(s): Irritant
Formula:
(C₆H₄F)OCH₂COOH MW: 170.14 g/mol Melting Pt: 106 °C Density: 1.516 g/cm³ (20 °C) |
MDL Number:
MFCD00004304 CAS Number: 405-79-8 EINECS: 206-974-5 |
Specification Test Results
Melting point | 104-105°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...