3-Fluoro-4-nitrobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

008960-5G 008960-25G
101795-940EA 56.92 USD
101795-940 101795-942
3-Fluoro-4-nitrobenzoic acid ≥97%
3-Fluoro-4-nitrobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄FNO₄
MW: 185.11
Melting Pt: 174…175 °C
MDL Number: MFCD01862092
CAS Number: 403-21-4

Order Now

Specification Test Results

Melting point 174-175°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR