2-Fluoro-6-methoxyphenol ≥98% for synthesis

Supplier: Matrix Scientific

008959-1G 008959-5G
101795-936EA 36.92 USD
101795-936 102832-800
2-Fluoro-6-methoxyphenol ≥98% for synthesis
2-Fluoro-6-methoxyphenol

Hazard code(s): Irritant

Formula: C₇H₇FO₂
Boiling Pt: 130…132 °C (36mm)
Density: 1.230 g/cm³
MDL Number: MFCD00075140
CAS Number: 73943-41-6

Order Now

Specification Test Results

Boiling point 130-132°C/3
Density 1.23
ND20 1.52


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR