6-Fluoro-o-anisaldehyde ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-6-mehoxybenzaldehyde, 2-Fluoro-6-methoxybenzaldehyde, 6-Fluoro-2-anisaldehyde

008955-1G 008955-5G 008955-25G
101795-922EA 16.92 USD
101795-922 101795-924 101795-926
6-Fluoro-o-anisaldehyde ≥95% for synthesis
6-Fluoro-o-anisaldehyde

Hazard code(s): Irritant, Store in Freezer, Air Sensitive

Formula: C₈H₇FO₂
Melting Pt: 59…63 °C
MDL Number: MFCD01090998
CAS Number: 146137-74-8

Order Now

Specification Test Results

Melting point 59-63°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR