4-Fluoro-2-iodotoluene ≥98% for synthesis

Supplier: Matrix Scientific
Danger

Synonyms: 1-Fluoro-3-iodo-4-methylbenzene, 4-Fluoro-2-iodo-1-methylbenzene

008953-5G 008953-25G 008953-100G
101795-910EA 20 USD
101795-910 101795-912 101795-914
4-Fluoro-2-iodotoluene ≥98% for synthesis
4-Fluoro-2-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆FI
Boiling Pt: 204 °C
MDL Number: MFCD00013709
CAS Number: 13194-67-7
EINECS: 236-153-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 204°C
ND20 1.578


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR