1-Bromo-2-chloro-3-fluorobenzene ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-3-fluorobromobenzene
008950-5G
008950-25G
008950-100G
101795-894EA
21.54
USD
101795-894
101795-896
101795-898
1-Bromo-2-chloro-3-fluorobenzene ≥98%
1-Bromo-2-chloro-3-fluorobenzene
Hazard code(s): Irritant
Formula:
C₆H₃BrClF MW: 209.44 g/mol |
MDL Number:
MFCD04038306 CAS Number: 883499-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...