1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

008933-1G 008933-5G
101795-850EA 69.23 USD
101795-850 101795-852
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate ≥97% for synthesis
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate

Hazard code(s): Irritant

Formula: C₇H₁₁F₃N₂O₃S
MDL Number: MFCD00210008
CAS Number: 145022-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR