2,3-Difluoro-4-hydroxybenzonitrile ≥97%
Supplier: Matrix Scientific
008922-1G
008922-5G
101795-828EA
60
USD
101795-828
101795-830
2,3-Difluoro-4-hydroxybenzonitrile ≥97%
2,3-Difluoro-4-hydroxybenzonitrile
Hazard code(s): Irritant
Formula:
C₇H₃F₂NO MW: 155.1 g/mol |
MDL Number:
MFCD00269596 CAS Number: 126162-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...