2,6-Dichloro-4-fluorophenol ≥97%

Supplier: Matrix Scientific
Warning

008898-25G 008898-100G
101795-756EA 109.23 USD
101795-756 101795-758
2,6-Dichloro-4-fluorophenol ≥97%
2,6-Dichloro-4-fluorophenol

Hazard code(s): Corrosive

Formula: C₆H₃Cl₂FO
MW: 180.99 g/mol
Boiling Pt: 142 °C (100mm)
Melting Pt: 49…51 °C
MDL Number: MFCD00010675
CAS Number: 392-71-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Boiling point 142°C/100mm
Melting point 49-51°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR