1,2-Dichloro-1,1-difluoroethane ≥97%

Supplier: Avantor

008894-25G
101795-740EA 63.25 USD
101795-740 101795-742
1,2-Dichloro-1,1-difluoroethane ≥97%
1,2-Dichloro-1,1-difluoroethane

Hazard code(s): Ozone Depleter, Irritant

Formula: ClF₂CCClH₂
Boiling Pt: 46…47 °C
Melting Pt: –101.2 °C
Density: 1.4163 g/cm³
MDL Number: MFCD00042125
CAS Number: 1649-08-7
EINECS: 216-714-2

Order Now

Specification Test Results

Boiling point 46-47°C
Melting point -101.2°C
Density 1.4163


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR