1-Chloro-1,1,2-trifluoro-2-iodoethane ≥97%
Supplier: Matrix Scientific
008875-5G
008875-25G
101795-688EA
261.54
USD
101795-688
101795-690
1-Chloro-1,1,2-trifluoro-2-iodoethane ≥97%
1-Chloro-1,1,2-trifluoro-2-iodoethane
Hazard code(s): Irritant
Formula:
C₂HClF₃I Boiling Pt: 77 °C (630mm) Density: 2.181 g/cm³ |
MDL Number:
MFCD00042138 CAS Number: 354-26-7 EINECS: 206-553-6 |
Specification Test Results
Boiling point | 77°C/630mm |
Density | 2.181 |
ND20 | 1.432 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...