2-Bromo-3-(trifluoromethyl)pyridine ≥98%
Supplier: Matrix Scientific
008858-5G
008858-25G
101795-640EA
20
USD
101795-640
101795-642
2-Bromo-3-(trifluoromethyl)pyridine ≥98%
2-Bromo-3-(trifluoromethyl)pyridine
Hazard code(s): Irritant
Formula:
C₆H₃BrF₃N MW: 226 g/mol Boiling Pt: 76 °C (8mm) Melting Pt: 37…39 °C |
MDL Number:
MFCD00153084 CAS Number: 175205-82-0 |
Specification Test Results
Boiling point | 76°C/8mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...