2'-Bromo-4'-fluoroacetanilide ≥97%

Supplier: Matrix Scientific

008837-5G 008837-25G
101795-568EA 47.69 USD
101795-568 101795-570
2'-Bromo-4'-fluoroacetanilide ≥97%
2'-Bromo-4'-fluoroacetanilide

Hazard code(s): Irritant

Formula: H₃CC(O)NH(C₆H₃BrF)
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 119 - 120 °C
MDL Number: MFCD00061118
CAS Number: 1009-22-9

Order Now

Specification Test Results

Melting point 117-119°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR