2-Hydroxy-5-nitropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Nitro-2-pyridinol

008780-25G 008780-100G
101795-440EA 29.23 USD
101795-440 101795-442
2-Hydroxy-5-nitropyridine ≥98%
2-Hydroxy-5-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Boiling Pt: 180…200 °C (0.2 mmHg)
Melting Pt: 188…191 °C
MDL Number: MFCD00006276
CAS Number: 5418-51-9
EINECS: 226-525-7

Order Now

Specification Test Results

Melting point 188-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR