2-Chloro-5-nitropyridine ≥98%

Supplier: Matrix Scientific
Warning

008779-25G 008779-100G
101795-436EA 21.54 USD
101795-436 101795-438
2-Chloro-5-nitropyridine ≥98%
2-Chloro-5-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00006240
CAS Number: 4548-45-2
EINECS: 224-908-3

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR