4-Benzyloxyindole ≥99%

Supplier: Matrix Scientific

008774-5G 008774-25G
101795-434EA 100 USD
101795-434 102832-798
4-Benzyloxyindole ≥99%
4-Benzyloxyindole

Hazard code(s): Irritant, Store in Freezer

Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Melting Pt: 59…61 °C
MDL Number: MFCD00047200
CAS Number: 20289-26-3
EINECS: 243-690-0

Order Now

Specification Test Results

Melting point 59-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR