4-Fluoro-2-nitrobenzoic acid ≥98%
Supplier: Matrix Scientific
008773-5G
008773-25G
008773-100G
101795-426EA
23.08
USD
101795-426
101795-428
101795-430
4-Fluoro-2-nitrobenzoic acid ≥98%
4-Fluoro-2-nitrobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄FNO₄ MW: 185.11 g/mol Melting Pt: 147…148 °C |
MDL Number:
MFCD00024300 CAS Number: 394-01-4 EINECS: 206-890-9 |
Specification Test Results
Melting point | 147-148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...