4-Bromothiophene-2-carbaldehyde ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde
008769-25G
008769-100G
101795-412EA
29.23
USD
101795-412
101795-414
4-Bromothiophene-2-carbaldehyde ≥98%
4-Bromothiophene-2-carbaldehyde
Hazard code(s): Irritant
Formula:
C₅H₃BrOS MW: 191.05 g/mol Boiling Pt: 114…115 °C (11mm) Melting Pt: 44…48 °C |
MDL Number:
MFCD00005431 CAS Number: 18791-75-8 EINECS: 242-577-3 |
Specification Test Results
Boiling point | 114-115°C/1 |
Melting point | 44-48°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...