4-Bromo-3-fluoroanisole ≥98% for synthesis

Supplier: Matrix Scientific
Warning

008761-5G 008761-25G 008761-100G
101795-382EA 24.62 USD
101795-382 101795-384 101795-386
4-Bromo-3-fluoroanisole ≥98% for synthesis
4-Bromo-3-fluoroanisole

Hazard code(s): Irritant

Formula: C₇H₆BrFO
MDL Number: MFCD01310983
CAS Number: 458-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR