2,3,6-Trifluoroisonicotinic acid ≥90%
Supplier: Matrix Scientific
008759-250MG
008759-1G
101795-376EA
73.85
USD
101795-376
101795-378
2,3,6-Trifluoroisonicotinic acid ≥90%
2,3,6-Trifluoroisonicotinic acid
Hazard code(s): Irritant
Formula:
C₆H₂F₃NO₂ Melting Pt: 125…127 °C |
MDL Number:
MFCD03840204 CAS Number: 675602-92-3 |
Specification Test Results
Melting point | 125-127°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...