2-Fluoro-6-hydroxybenzonitrile ≥98%

Supplier: Matrix Scientific

008753-1G 008753-5G
101795-360EA 16.92 USD
101795-360 101795-362
2-Fluoro-6-hydroxybenzonitrile ≥98%
2-Fluoro-6-hydroxybenzonitrile

Hazard code(s): Irritant

Formula: C₇H₄FNO
MW: 137.11 g/mol
Melting Pt: 155…157 °C
MDL Number: MFCD03428592
CAS Number: 140675-43-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR