7-Fluoroisatin ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 7-Fluoro-1H-indole-2,3-dione, 7-Fluoroindoline-2,3-dione

008742-5G 008742-25G 008742-100G
101795-338EA 15.38 USD
101795-338 101795-340 101795-342
7-Fluoroisatin ≥97%
7-Fluoroisatin

Hazard code(s): Irritant

Formula: C₈H₄FNO₂
MW: 165.12 g/mol
MDL Number: MFCD01569508
CAS Number: 317-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR