3-Fluoro-p-anisidine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-anisidine, 3-Fluoro-4-methoxyaniline, 4-Amino-2-fluoroanisole

008741-5G 008741-25G
101795-334EA 16.92 USD
101795-334 101795-336
3-Fluoro-p-anisidine ≥98%
3-Fluoro-p-anisidine

Hazard code(s): Irritant

Formula: C₇H₈FNO
MW: 141.15 g/mol
Melting Pt: 81…83 °C
MDL Number: MFCD00075040
CAS Number: 366-99-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR