2-Chloro-4-nitrobenzotrifluoride ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-nitro-1-(trifluoromethyl)benzene, 2-Chloro-4-nitrobenzotrifluoride, 1-Chloro-3-nitro-6-(trifluoromethyl)benzene

008737-1G 008737-5G 008737-25G
101795-324EA 21.54 USD
101795-324 101795-326 101795-328
2-Chloro-4-nitrobenzotrifluoride ≥98%
2-Chloro-4-nitrobenzotrifluoride

Hazard code(s): Irritant

Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
MDL Number: MFCD00171532
CAS Number: 151504-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR