3-Chloro-4-(trifluoromethyl)aniline ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-chlorobenzotrifluoride, 3-Chloro-α,α,α-trifluoro-p-toluidine, 4-Amino-2-chloro-α,α,α-trifluortoluene, 4-Amino-2-chloro-α,α,α-trifluorotoluene

008733-5G 008733-25G
101795-314EA 64.62 USD
101795-314 101795-316
3-Chloro-4-(trifluoromethyl)aniline ≥97% for synthesis
3-Chloro-4-(trifluoromethyl)aniline

Hazard code(s): Irritant

Formula: C₇H₅ClF₃N
MDL Number: MFCD00278479
CAS Number: 445-13-6
EINECS: 207-152-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR