3-Methyl-5-(trifluoromethyl)-1H-pyrazole ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Methyl-5-(trifluoromethyl)pyrazole

008727-5G 008727-25G
101795-304EA 44.62 USD
101795-304 101795-306
3-Methyl-5-(trifluoromethyl)-1H-pyrazole ≥98%
3-Methyl-5-(trifluoromethyl)-1H-pyrazole

Hazard code(s): Irritant

Formula: C₅H₅F₃N₂
MW: 150.104 g/mol
Melting Pt: 89…90 °C
MDL Number: MFCD00829309
CAS Number: 10010-93-2

Order Now

Specification Test Results

Melting point 89-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR