4-Pyridylmethanol ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Pyridylmethanol, 4-(Hydroxymethyl)pyridine, 4-Pyridinemethanol, Pyridine-4-methanol, 4-Pyridylcarbinol

008724-25G 008724-100G
101795-292EA 29.23 USD
101795-292 101795-294
4-Pyridylmethanol ≥98%
4-Pyridylmethanol

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 141 °C (12mm)
Melting Pt: 53…55 °C
Density: 1.15 g/cm³ (20 °C)
MDL Number: MFCD00006442
CAS Number: 586-95-8
EINECS: 209-590-6

Order Now

Specification Test Results

Boiling point 141°C/12mm
Melting point 53-55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR