6-Hydroxynicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid

008720-25G 008720-100G
101795-284EA 26.15 USD
101795-284 101795-286
6-Hydroxynicotinic acid ≥98%
6-Hydroxynicotinic acid

Hazard code(s): Irritant

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 304 - 306 °C
MDL Number: MFCD00006277
CAS Number: 5006-66-6
EINECS: 225-682-9

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR