4,4'-Diiodobiphenyl ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 4,4'-Diiodo-1,1'-biphenyl

008711-25G 008711-100G
101795-256EA 21.54 USD
101795-256 101795-258
4,4'-Diiodobiphenyl ≥98% for synthesis
4,4'-Diiodobiphenyl

Hazard code(s): Irritant, Light Sensitive

Formula: C₁₂H₈I₂
MW: 406 g/mol
Melting Pt: 201…204 °C
MDL Number: MFCD00001057
CAS Number: 3001-15-8
EINECS: 221-080-5

Order Now

Specification Test Results

Melting point 201-204°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR