4-Iodobiphenyl ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Iodo-1,1'-biphenyl

008710-5G 008710-25G
101795-252EA 44.62 USD
101795-252 101795-254
4-Iodobiphenyl ≥97% for synthesis
4-Iodobiphenyl

Hazard code(s): Irritant, Light Sensitive

Formula: C₁₂H₉I
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 110…114 °C
Density: 1.584 g/cm³ (20 °C)
MDL Number: MFCD00019028
CAS Number: 1591-31-7
EINECS: 216-469-1

Order Now

Specification Test Results

Melting point 110-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR