6-(Trifluoromethyl)uracil ≥99% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)uracil, 2,4-Dihydroxy-6-trifluoromethylpyrimidine

008709-1G 008709-5G 008709-25G
101795-246EA 33.85 USD
101795-246 101795-248 101795-250
6-(Trifluoromethyl)uracil ≥99% for synthesis
6-(Trifluoromethyl)uracil

Hazard code(s): Irritant

Formula: C₅H₃F₃N₂O₂
MW: 180.09 g/mol
Melting Pt: 229…230 °C
MDL Number: MFCD01011762
CAS Number: 672-45-7

Order Now

Specification Test Results

Melting point 229-230°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR