4-Bromo-3-fluorophenol ≥97%

Supplier: Matrix Scientific

008707-25G 008707-100G
101795-238EA 72.31 USD
101795-238 101795-240
4-Bromo-3-fluorophenol ≥97%
4-Bromo-3-fluorophenol

Hazard code(s): Irritant

Formula: C₆H₄BrFO
MW: 191
Melting Pt: 71…72 °C
MDL Number: MFCD00051907
CAS Number: 121219-03-2

Order Now

Specification Test Results

Melting point 71-72°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR