2-Phenylpyridine ≥98%

Supplier: Matrix Scientific
Warning

008704-25G 008704-100G
101795-224EA 44.62 USD
101795-224 101795-226
2-Phenylpyridine ≥98%
2-Phenylpyridine

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 269 - 270 °C (1013 hPa)
Melting Pt: 76 - 77 °C
Density: 1.623 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006280
CAS Number: 1008-89-5
EINECS: 213-763-1
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 268-270°C
Density 1.086
ND20 1.623

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR