4-Iodophenol ≥98%

Supplier: Matrix Scientific

008702-25G 008702-100G
101795-220EA 84.62 USD
101795-220 101795-222
4-Iodophenol ≥98%
4-Iodophenol

Hazard code(s): Irritant, Lachrymator

Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 92 - 93 °C
Density: 2.13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002327
CAS Number: 540-38-5
EINECS: 208-745-5
UN: 3261
ADR: 8,III
Merck Index: 13,05058

Order Now

Specification Test Results

Melting point 92-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR