2-Iodophenol ≥98% for synthesis

Supplier: Matrix Scientific
Warning

008701-25G 008701-100G
101795-216EA 58.46 USD
101795-216 101795-218
2-Iodophenol ≥98% for synthesis
2-Iodophenol

Hazard code(s): Irritant, Light Sensitive

Formula: C₆H₅IO
Boiling Pt: 186 - 187 °C (213 hPa)
Melting Pt: 39 - 42 °C
Density: 1.8757 g/cm³ (80 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013963
CAS Number: 533-58-4
EINECS: 208-569-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,05057

Order Now

Specification Test Results

Boiling point 186-187°C/1
Melting point 39-41°C
Density 1.947


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR