4-Chloro-2-fluorophenol ≥97%

Supplier: Matrix Scientific

008696-25G 008696-100G
101795-202EA 52.31 USD
101795-202 101795-204
4-Chloro-2-fluorophenol ≥97%
4-Chloro-2-fluorophenol

Hazard code(s): Irritant

Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 103…104 °C (50mm)
Melting Pt: 20 °C
MDL Number: MFCD00075030
CAS Number: 348-62-9

Order Now

Specification Test Results

Boiling point 103-104°C/5
Melting point 20°C
ND20 1.536

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR