2,4,5-Trifluoro-4-hydroxybenzoic acid ≥99%

Supplier: Matrix Scientific

008695-25G 008695-100G 008695-500G
101795-196EA 23.08 USD
101795-196 101795-198 101795-200
2,4,5-Trifluoro-4-hydroxybenzoic acid ≥99%
2,4,5-Trifluoro-4-hydroxybenzoic acid

Hazard code(s): Irritant

Formula: C₇H₃F₃O₃
MDL Number: MFCD00800378
CAS Number: 116751-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR