2-Bromo-3,5-difluoropyridine ≥98%
Supplier: Matrix Scientific
008691-1G
008691-5G
008691-25G
101795-180EA
73.85
USD
101795-180
101795-182
101795-184
2-Bromo-3,5-difluoropyridine ≥98%
2-Bromo-3,5-difluoropyridine
Hazard code(s): Irritant
Formula:
C₅H₂BrF₂N Boiling Pt: 36…37 °C (0.6mm) |
MDL Number:
MFCD03840197 CAS Number: 660425-16-1 |
Specification Test Results
Boiling point | 36-37°C/0.6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...