(±)-Nipecotic acid ≥98%

Supplier: Matrix Scientific

Synonyms: H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid

008689-25G 008689-100G
101795-176EA 16.92 USD
101795-176 101795-178
(±)-Nipecotic acid ≥98%
(±)-Nipecotic acid

Hazard code(s): Irritant

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: ∼261 °C (dec.)
MDL Number: MFCD00005992
CAS Number: 498-95-3
EINECS: 207-873-9

Order Now

Specification Test Results

Melting point ca 261°C(de

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR