Thiophene-3-carbaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde

008686-25G 008686-100G
101795-172EA 38.46 USD
101795-172 101795-174
Thiophene-3-carbaldehyde ≥98%
Thiophene-3-carbaldehyde

Hazard code(s): Air Sensitive, Refrigerate

Formula: C₅H₄OS
MW: 112.15 g/mol
Boiling Pt: 191 °C (1013 hPa)
Melting Pt: -30 °C
Density: 1.28 g/cm³ (24 °C)
MDL Number: MFCD00005466
CAS Number: 498-62-4
EINECS: 207-865-5

Order Now

Specification Test Results

Boiling point 80-81°C/14m
Density 1.228
ND20 1.583

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR