3-Amino-4-fluorobenzoic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Amino-4-fluorobenzoic Acid
008683-5G
008683-25G
008683-100G
101795-162EA
355.38
USD
101795-162
101795-164
101795-166
3-Amino-4-fluorobenzoic acid ≥97%
3-Amino-4-fluorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol |
MDL Number:
MFCD00130055 CAS Number: 2365-85-7 EINECS: 219-122-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...