6-Methylindole tech for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 6-Methyl-1H-indole

008681-1G 008681-5G
101795-158EA 15.38 USD
101795-158 101795-160
6-Methylindole tech for synthesis
6-Methylindole

Hazard code(s): Irritant, Light Sensitive, Store in Freezer

Formula: C₉H₉N
Boiling Pt: 112 °C (5mm)
Melting Pt: 29…32 °C
Density: 1.05 g/cm³ (20 °C)
MDL Number: MFCD00005682
CAS Number: 3420-02-8
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Boiling point 112°C/5mm
Melting point 29-32°C
Density 1.059
ND20 1.6071


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR