Indole-5-carbonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole

008677-5G 008677-25G 008677-100G
101795-152EA 33.85 USD
101795-152 101795-154 101795-156
Indole-5-carbonitrile ≥98%
Indole-5-carbonitrile

Hazard code(s): Irritant-Harmful, Air Sensitive, Store in Freezer, Light Sensitive

Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD00005669
CAS Number: 15861-24-2
EINECS: 239-986-4

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR