5-Hydroxynicotinic acid ≥97%

Supplier: Matrix Scientific

008183-25G 008183-100G
101794-188EA 69.23 USD
101794-188 102663-570
5-Hydroxynicotinic acid ≥97%
5-Hydroxynicotinic acid

Hazard code(s): Irritant

Formula: C₆H₅NO₃
MDL Number: MFCD00129117
CAS Number: 27828-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR