1-Propylboronic acid for synthesis

Supplier: Matrix Scientific

Synonyms: Propylboronic acid

007707-1G 007707-5G
101793-260EA 15.38 USD
101793-260 101793-262
1-Propylboronic acid for synthesis
1-Propylboronic acid
Formula: C₃H₉BO₂
MDL Number: MFCD01074564
CAS Number: 17745-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR