4-(Bromomethyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (4-(Bromomethyl)phenyl)metaboric acid, (4-(Bromomethyl)phenyl)boronic acid, Dihydroxy(4-(bromomethyl)phenyl)borane, 4-Bromomethylphenylboronic acid, 4-(Bromomethyl)benzeneboronic acid

007675-1G 007675-5G
101793-216EA 20 USD
101793-216 101793-218
4-(Bromomethyl)phenylboronic acid ≥97%
4-(Bromomethyl)phenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
MDL Number: MFCD01318113
CAS Number: 68162-47-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR